missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthene, 97%

Product Code. 15483097
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 15483097

Brand: Thermo Scientific Alfa Aesar A14922.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 83-32-9 | C12H10 | 154.21 g/mol

Acenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.

Solubility
Soluble in chloroform, benzene, tolune, glacial acetic acid and methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and alkaline earth metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-32-9
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003807
InChI Key CWRYPZZKDGJXCA-UHFFFAOYSA-N
Synonym acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y
PubChem CID 6734
ChEBI CHEBI:22154
IUPAC Name 1,2-dihydroacenaphthylene
SMILES C1CC2=C3C1=CC=CC3=CC=C2

Specifications

Melting Point 92°C to 95°C
Boiling Point 278°C
Flash Point 135°C (275°F)
Quantity 50 g
UN Number UN3077
Beilstein 386081
Merck Index 14,28
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material Acenaphthene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.