Learn More
Acebilustat, MedChemExpress
Product Code. 16477428
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16477428
Brand: Medchem Express HY17625
MedChemExpress Acebilustat (CTX-4430) is a leukotriene A4 hydrolase inhibitor, used for an oral antiinflammatory drug.
Acebilustat (CTX-4430) is a leukotriene A4 hydrolase inhibitor, used for an oral antiinflammatory drug.
Specifications
Chemical Name or Material Acebilustat
CAS 943764-99-6
Color White
Purity Grade Notes Research
Molecular Formula C29H27N3O4
Quantity 10 mg
Synonym CTX-4430
Solubility Information DMSO : 16.67 mg/mL (34.62 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(CN2[C@](C3)([H])CN(CC4=CC=C(OC5=CC=C(C6=NC=CO6)C=C5)C=C4)[C@]3([H])C2)C=C1
Molecular Weight (g/mol) 481.54
Formula Weight 481.54
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.