Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
Brand:  Fisherbrand™
Additional Details : Weight : 2.60000kg
 View more versions of this product
Product Code. 15489709
  • 9450.00 NOK / Each
Estimated Shipment: 18-10-2024
Description
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
Specifications
Accu100
AccuQuatro U pack Cartridge Kit with 0.2μm final filter, RO feed