Learn More
Thermo Scientific™ Accucore™ C18 LC Column
Increase sample throughput and boost laboratory productivity using Thermo Scientific™ Accucore™ C18 LC Columns.
Brand:  Thermo Scientific™ 17126-101030
Additional Details : Weight : 10.00000kg
Product Code. 13400517
  • 10900.68 NOK / Each
Estimated Shipment: 14-05-2024
Description
  • Optimal Selectivity for Nonpolar Compounds
  • Hydrophobic interaction mechanism
  • Fast Separations
  • 2.6μm core-enhanced technology particles enable operation at high flow rates without generating excessive backpressures.
  • Excellent Peak Shapes for Enhanced Resolution and Sensitivity
  • Solid core 2.6μm particles and narrow particle size distribution reduce band broadening and improve separation efficiencies.
  • High surface coverage of silica minimizes secondary interactions and peak tailing.
  • Low Backpressures
  • Solid core 2.6μm particles for less backpressure compared to UHPLC sub-2μm particles
  • Compatible with conventional HPLC instrumentation and UHPLC platforms
  • Excellent Reproducibility and Robustness
  • Advanced bonding technology
  • Automated packing process
Specifications
Specifications
100 mm
2.6 μm