Learn More
Fisher Scientific™ Accu500 Water System
Shop All Fisherbrand Products
Click to view available options
Type:
UV Ultrapure Water System
UV Ultrapure Water System w/ Dispenser
Ultrapure Water Purification System
RO Flow Rate:
10 L/hr.
15 L/hr.
15 L/min.
20 L/hr.
30 L/hr.
Unit Size:
Each
Beskrivning
- The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
- The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
- Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
- Automatic hourly system recirculation prevents bacterial growth
- Resistivity is measured by a highly sensitive resistivity sensor
- Electrical and water compartments are completely segregated, thus safer to operate
- User can set water dispensing time for easy water collection
- Both RO and ultrapure water can be dispensed from the water system
- The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
- A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
- Independent ultrapure water dispenser
- The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
- Volumetric dispensing and calibration
- The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:
Ultrapure water
- HPLC mobile phase preparation
- Preparation of reagent blank solutions
- As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
- Preparation of buffers and culture media for mammalian cell culture
- Preparation of molecular biology reagents, etc.
RO water
- Glassware cleaning
- Washing machine for glassware
- Waterbath water
- Autoclaving
Specifikationer
Specifikationer
| Conductivity | <20μS/cm |
| Endotoxin Concentration | <0.001 EU/mL with a terminal ultrafiltration cartridge |
| Product Water Type | Tap Water |
| Resistivity | 18.2 Mohm.cm |
| Type | UV Ultrapure Water System |
| Feedwater Pressure | 1 to 6 bar |
| Model | Accu500 |
| Particles >0.2µM/mL | <1/mL with 0.2μm final filter or terminal ultrafiltration cartridge |
| Purified Water Flowrate Field | 1.5 L/min. |
| RO Flow Rate | 10 L/hr. |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering