Learn More
Fisherbrand™ ACCU500 Water Purification System
Product Code. 15489699 Shop All Fisherbrand Products
Product Code. 15489699
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Produces ultrapure and reverse osmosis water from tap water directly with no additional pre-treatment filters.
  • Quality of ultrapure water meets or exceeds ASTM, CLSI, CAP and ISO Type I water standards
  • System CE and RoHS certified
  • Remote dispenser with built-in high precision resistivity sensor with cell constant of 0.01 cm-1 to measure ultrapure water quality just prior to the dispensing point
  • Two years warranty
Specifications
Certifications/Compliance ASTM, CLSI, CAP, ISO Type I
Conductivity <20µS/cm
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Feedwater Source Tap, TDS < 1000 ppm (2000μS/cm)
Flow Rate 1.5 L/min. (ultrapure water type I)
Product Water Type Type 1 and 3
Remote Dispenser Optional
Resistivity 18.2 Mohm.cm
Particles >0.2µM/mL <1/mL (with a 0.2μm final filter or terminal ultrafiltration cartridge)
RO Flow Rate 10, 15, 20, 30 L/hr.
Dimensions (L x W x H) 30 x 51 x 48 cm