Learn More
Brand™ accu-jet™ pro Pipet Controller
Shop All Brand Products
Click to view available options
Color:
Dark Blue
Green
Magenta
Royal Blue
Plug Type:
Europe (continent)
N/A - charger not included
Unit Size:
Each
Description
- One-handed delivery mode and selection
- Ergonomic design is perfectly balanced, weighing only 190g for fatigue-free pipetting even in prolonged operations.
- Adjustable motor speed limiter – a slower speed setting allows even more sensitive control of filling and dispensing rates for lower volume pipets.
- A flashing LED indicates 2 hours in advance when the battery needs charging. Approximately eight hours of continuous pipetting (with a 10mL pipet) without recharging.
- Direct outlet of liquid vapors protects against corrosion to ensure long instrument life.
- A 0.2μm hydrophobic membrane filter and a built-in check valve protect against liquid penetrating inside. Perfect fit for glass and plastic pipets from 0.1 to 200mL
Specifications
Specifications
| Battery Type | Nickel-metal hydride |
| Color | Dark Blue |
| Pore Size | 0.2 μm |
| Type | Pipette Controller |
| Filter Type | Hydrophobic membrane |
| Includes | Nickel-metal hydride battery, 2 battery compartment covers, wall support, AC adapter (100-240V; 50/60Hz) and 2 spare membrane filters 0.2µm, sterile. 26304 without recharger |
| Plug Type | Europe (continent) |
| Quantity | 1 Each |
| Weight (Metric) | 190 g |
| Voltage | 100/240 V |
| Show More |