Learn More
Thermo Scientific™ Acclaim™ 120 C18 Columns
Shop All Thermo Scientific ProductsDescription
Improve the separation efficiency in your reversed-phase applications with Thermo Scientific™ Acclaim™ 120 C18 columns. These columns feature an ultrapure silica substrate with extremely low metal content to minimize tailing effects and deliver symmetrical peak shapes. The unique bonding chemistry yields excellent surface coverage to provide highly predictable separations that are unaffected by secondary interactions. Use these high-efficiency columns for maximum resolution in a wide variety of applications.
- Very high surface coverage and very low metal content together result in columns with excellent efficiencies
- Reproducible column-to-column performance
- Extremely low bleed, fully compatible with MS
- Stable between pH 2 and 8
- Available in 2.2, 3, and 5ÎĽm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 120Ă…
- Also available in 2.2ÎĽm RSLC format for reliable, fast separations
Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.
- Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
- Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Specifications
Specifications
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 50 mm |
Max. Pressure | 4500 psi (310 bar) |
Particle Size | 5 ÎĽm |
pH | 2 to 8 |
Pore Size | 120 Ă… |
Quantity | Each |
Surface Area | 300 m²/g |
Temperature | 60°C |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Documents
Scientific Resources
Application Notes
Posters
Brochures
Flyers
Product Information
Manuals
Specification Sheets