Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Product Code. 10767381 Shop All IKA Products
Change view
Click to view available options
Model:
R 1300
R 1302
R 1311
R 1342
R 1345
R 1352
R 1375
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10767381 R 1300 Each
10568881 R 1375 Each
10201961 R 1302 Each
10160161 R 1311 Each
10642781 R 1345 Each
10058130 R 1352 Each
10435261 R 1342 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10767381 Supplier IKA™ Supplier No. 0000513500
  • ONLINE EXCLUSIVE PRICE
    4000.00 NOK
    3904.27 NOK / Pack of 1
    Save 95.73 NOK
    2% Off
In Stock
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
  • High turbulence, high shearing forces
  • Particle reduction
  • Used at medium to high speeds
† PTFE-coated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 3.1 in.
Diameter (English) Shaft 0.3 in.
Diameter (Metric) 80 mm
Diameter (Metric) Shaft 8 mm
Length (English) 13.8 in.
Length (Metric) 350 mm
Model R 1300
Speed 2000 rpm max.
Type Dissolver Stirrer
Material Stainless Steel (316L)