Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Product Code. 10201961 Shop All IKA Products
Change view
Click to view available options
Model:
R 1300
R 1302
R 1311
R 1342
R 1345
R 1352
R 1375
Unit Size:
Each
Product Code. Model unitSize
10201961 R 1302 Each
10568881 R 1375 Each
10767381 R 1300 Each
10160161 R 1311 Each
10642781 R 1345 Each
10058130 R 1352 Each
10435261 R 1342 Each
7 options
Product Code. 10201961
Brand: IKA™ 0002387900
Only null left
Add to basket
Add to basket
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
  • High turbulence, high shearing forces
  • Particle reduction
  • Used at medium to high speeds
† PTFE-coated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 3.9 in.
Diameter (English) Shaft 0.4 in.
Diameter (Metric) 100 mm
Diameter (Metric) Shaft 10 mm
Length (English) 13.8 in.
Length (Metric) 350 mm
Model R 1302
Speed 1000 rpm max.
Type Dissolver Stirrer
Material Stainless Steel (316L)