Learn More
IKA™ R 1303 Dissolver Stirrer
Product Code. 10005220 Shop All IKA Products
Change view
Click to view available options
Length (Metric):
350 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10005220 350 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10005220 Supplier IKA™ Supplier No. 0002746700
  • ONLINE EXCLUSIVE PRICE
    6090.00 NOK
    5240.39 NOK / Pack of 1
    Save 849.61 NOK
    13% Off
In Stock
Draws material to be mixed from the top and the bottom with radial flow. IKA R 1303 Dissolver Stirrer creates high turbulence and high shearing forces with particle reduction. For use at medium to high speeds.
Radial flow, for drawing the material to be mixed from the top and the bottom. High turbulence, high shearing forces. Particle reduction. Used at medium to high speeds. Stirrer Ø: 42 mm Shaft Ø: 8 mm Shaft length: 350 mm
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Fan 1.7 in.
Diameter (English) Shaft 0.3 in.
Diameter (Metric) Fan 42 mm
Diameter (Metric) Shaft 8 mm
Length (English) 13.8 in.
Length (Metric) 350 mm
Speed 2000 rpm
Type Stirring Shaft