Learn More
Thermo Scientific™ Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Product Code. 15640845 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Liquid Filter
Storage Bottle Cap
For Use With (Equipment):
Wellwash Microplate Strip Washer
Unit Size:
1 set
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
15640845 Liquid Filter Wellwash Microplate Strip Washer 1 set
15331441 Storage Bottle Cap Wellwash Microplate Strip Washer Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15640845 Supplier Thermo Scientific™ Supplier No. N12407
Estimated Shipment: 22-12-2026
Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Pk.
For Use With (Equipment) Wellwash Microplate Strip Washer
Product Type Liquid Filter