Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Product Code. 10688878 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Adapter
Adjustable Bridge
Bridge
Cage
Extension Bridge
Female M16x1 Adapter (NPTF)
Fluid Displacement Block
Lifting Platform
Male M16x1 Adapter (NPTF)
Tap Water Cooling Coil
Trolley with Castors
For Use With (Equipment):
AC150 or AC200 controller with S3
Cold/Heated Bath Circulators
For A10B
For A25 or A40
For A28 or A25
For A40
For A5B
For A5B, A10B liftplate
For S15 Heated Bath Circulator
For S15, S21, S30, S45
For S21, S30, S45
For S49 Heated Bath Circulator
RS232 Cable for Bath Circulator
S15, S21, S30, S45 Liftplate
S49 Liftplate
SC Controller on Water Baths
SC, AC, PC immersion Circulators
SC100 or SC150 Immersion Circulator
SC100, SC150, AC150, AC200 , PC200
SC100/SC150 Circulator with S3
SC100/SC150 Circulator with S5P
SC100/SC150 Circulator with S6T
SC150L Circulator with S7
SC150L Immersion Circulator
SC150L Immersion Circulator+clamp
SC150L with S13, S15, S30, S45, S49
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
10688878 Adjustable Bridge SC, AC, PC immersion Circulators Each
11660961 Extension Bridge S15, S21, S30, S45 Liftplate Each
11680961 Extension Bridge S49 Liftplate Each
10638845 Lifting Platform For S21, S30, S45 Each
11610971 Lifting Platform For S15 Heated Bath Circulator Each
15150965 Lifting Platform For S49 Heated Bath Circulator Each
11620971 Lifting Platform For S49 Heated Bath Circulator Each
10030638 Tap Water Cooling Coil SC100, SC150, AC150, AC200 , PC200 Each
11640971 Tap Water Cooling Coil SC100 or SC150 Immersion Circulator Each
11650971 Tap Water Cooling Coil SC150L with S13, S15, S30, S45, S49 Each
11660971 Tap Water Cooling Coil SC150L Immersion Circulator+clamp Each
10419204 Adapter RS232 Cable for Bath Circulator Each
11697998 Female M16x1 Adapter (NPTF) Cold/Heated Bath Circulators Each
11931276 Male M16x1 Adapter (NPTF) Cold/Heated Bath Circulators Each
10393705 Extension Bridge For A5B, A10B liftplate Each
11901286 Trolley with Castors For A40 Each
11911286 Trolley with Castors For A28 or A25 Each
11670961 Extension Bridge For S15, S21, S30, S45 Each
10332295 Bridge SC Controller on Water Baths Each
11951286 Cage SC100 or SC150 Immersion Circulator Each
11961286 Cage SC150L Immersion Circulator Each
11618018 Tap Water Cooling Coil SC100/SC150 Circulator with S5P Each
11628018 Tap Water Cooling Coil SC100/SC150 Circulator with S6T Each
11630833 Tap Water Cooling Coil SC100/SC150 Circulator with S3 Each
11901296 Tap Water Cooling Coil SC150L Circulator with S7 Each
10324078 Tap Water Cooling Coil AC150 or AC200 controller with S3 Each
15474543 Fluid Displacement Block For A25 or A40 Each
15321767 Lifting Platform For A5B Each
13433836 Lifting Platform For A10B Each
29 options
Product Code. 10688878
Brand: Thermo Scientific™ 1600018
Only null left
Add to basket
Add to basket
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SC, AC, PC immersion Circulators
Type Adjustable Bridge
Width (Metric) Top Plate 300 to 800 mm