Learn More
Thermo Scientific™ Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Product Code. 15228904 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
170 mm
220 mm
Type:
Plate Cover
Unit Size:
Each
Product Code. 15228904
Brand: Thermo Scientific™ 88880145
Only null left
Add to basket
Add to basket
Use these accessories to optimize performance of your Thermo Scientific™ RT Basic Series Magnetic Stirrers.
Use these Thermo Scientific™ Accessories for RT Basic Series Magnetic Stirrers to optimize performance of your stirrer.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Diameter (Metric) 170 mm
For Use With (Equipment) RT Basic Series and RT Touch Series Magnetic Stirrers
Holds 96-Well Plates, 384-Well Plates
Type Plate Cover
Material Silicone