Learn More
Sartorius Accessories and Replacement Parts for Balances: Switches
GREENER_CHOICE
Product Code. 10049510 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
CPA Balances, Extended Balances, Master Pro LA Balances
Draft Shield of SE and ME Ultramicro and Microbalance
Type:
Foot Switch
Hand Switch
Unit Size:
Each
Product Code. For Use With (Equipment) Type unitSize
10049510 Draft Shield of SE and ME Ultramicro and Microbalance Foot Switch Each
10062221 CPA Balances, Extended Balances, Master Pro LA Balances Hand Switch Each
10110751 CPA Balances, Extended Balances, Master Pro LA Balances Foot Switch Each
3 options
Product Code. 10049510
Brand: Sartorius YPE01RC
Balance Switches for remote control.
  • Hand and foot switch accessories for Sartorius balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Draft Shield of SE and ME Ultramicro and Microbalance
Includes T-connector
Type Foot Switch