Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Product Code. 12043273
Change view
Click to view available options
Type:
Silicone Oil
Vial Cleaning Brushes
Vials
Unit Size:
Each
Pack of 10
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12043273 Vial Cleaning Brushes Pack of 10
12033273 Silicone Oil Each
12023273 Vials Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12043273 Supplier Thermo Scientific™ Supplier No. AC3CBR
  • ONLINE EXCLUSIVE PRICE
    390.00 NOK
    363.83 NOK / Pack of 10
    Save 26.17 NOK
    6% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Features:
  • Accessories for AQUAfast AQ3010 Turbidity Meter
TRUSTED_SUSTAINABILITY
Specifications
Type Vial Cleaning Brushes
For Use With (Equipment) AQUAfast AQ3010 Turbidity Meter
Description Vial Cleaning Brushes, 5 Pack