Learn More
Cytiva Accessories for Multiphor™ II Electrophoresis System
Shop All Cytiva Products
Click to view available options
Type:
EPH Electrode Anode
EPH Electrode Cathode
EPH Electrode Wick (104 x 253 mm)
EPH Electrode Wick (82 x 130 mm)
Flexiclamp
IEF Electrode Strip
Unit Size:
Each
Pack of 100
Pack of 500
Pack of 6
Specifications
Specifications
| For Use With (Application) | Standard flatbed unit |
| For Use With (Equipment) | Multiphor II Electrophoresis System |
| Type | EPH Electrode Wick (104 x 253 mm) |