Description
Accommodate various applications and budgets with these flexible and convenient Thermo Scientific™ MaxQ™ 8000 Shaker Accessories. With exceptional simplicity and flexibility, these shaker accessories are the optimal choice for busy labs.
Specifications
Specifications
| Product Type | Stand for 2 Stackable Shakers |
| Depth (English) | 30.5 in. |
| Depth (Metric) | 75.2 cm |
| Height (English) | 11.8 in. |
| Height (Metric) | 30 cm |
| Width (English) | 45.5 in. |
| Width (Metric) | 115.6 cm |
| Weight (English) | 142 lb. |
| For Use With (Equipment) | MaxQ 8000 |