Learn More
Thermo Scientific™ Isotemp™ Gas Tank Switcher
Product Code. 11892711 Shop All Thermo Scientific Products
Click to view available options
Voltage:
120 V
Unit Size:
Each
Product Code. 11892711
missing translation for 'mfr': Thermo Scientific™ 3050
Only null left
Add to basket
Add to basket
For Use With: Napco 8000; Water Jacket Models Only
  • Napco 8000; Water Jacket Models Only
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Napco 8000; Water Jacket Models Only
Type Gas Tank Switcher
Electrical Requirements 120 V, 60 Hz
Voltage 120 V
Frequency 60 Hz