Learn More
Cytiva Accessories For Multiphor™ II Electrophoresis Systems
Product Code. 11560824 Shop All Cytiva Products
Click to view available options
Type:
Mylar Sheet
Sample Application Piece
Unit Size:
Pack of 200
Pack of 50
Product Code. 11560824
Brand: Cytiva 1850901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessories for analytical or preparative separations of proteins and peptides by isoelectric focusing (IEF), native or SDS PAGE, 2-D PAGE and DNA electrophoresis
  • Assists high quality production of lab-cast gels
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Sample Application Piece
Quantity 200/Pk