Learn More
Fisherbrand™ Accessories for Fisherbrand™ RT Basic Series Magnetic Stirrers
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
170 mm
220 mm
missing translation for 'unitSize'
Each
Description
- White
- Silicone
Specifications
Specifications
| Diameter (Metric) | 170 mm |
| For Use With (Equipment) | RT Basic Series Magnetic Stirrer |
| Type | White Silicone Plate Cover |