Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Product Code. 15564090 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
1.5/2.0 mL Tubes
10-15 mL and 5-7 mL Tubes
14 mm dia. Culture Tubes
15 mL Tubes
18.5 mm dia. Culture Tubes
50 mL Tubes
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15564090 15 mL Tubes Each
15554090 1.5/2.0 mL Tubes Each
15574090 50 mL Tubes Each
15584090 10-15 mL and 5-7 mL Tubes Each
15594090 14 mm dia. Culture Tubes Each
15504100 18.5 mm dia. Culture Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15564090
  • ONLINE EXCLUSIVE PRICE
    1895.00 NOK
    1426.93 NOK / Pack of 1
    Save 468.07 NOK
    24% Off
Estimated Shipment: 22-12-2026
Optional accessories to enhance the Fisherbrand™Multi-Purpose Tube Rotator
Multi-purpose tube rotator offers flexibility with multiple carousel options
  • Tube rotator
TRUSTED_SUSTAINABILITY
Specifications
Holds 15 mL Tubes
Product Type Tube Carousel
No. of Positions 30