Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker
Shop All Fisherbrand Products
Click to view available options
Product Type:
Adjustable Tube Rack
Fixed Tube Rack
Flask Clamp
Funnel Clamp
Infusion Bottle Clamp
Platform with Adjustable Bars
Platform with Clamps
Platform with Rubber Mat
Holds:
1000 mL Flask
1000 mL Infustion Bottle
12 x 100 mL Flasks, 150 mL Beakers
2000 mL Flask
3000 mL Flask
40 x 14 mm dia. Tubes
40 x 16 mm dia. Tubes
500 mL Flask
500 mL Infusion Bottle
5000 mL
6 x 250 mL Flasks, 150 mL Beakers
Funnels
Vessels
Unit Size:
Each
Description
Accessories for multi platform shaker to enhance performance and flexibility
- Increase performance and flexibility
Specifications
Specifications
| Holds | 40 x 14 mm dia. Tubes |
| Product Type | Fixed Tube Rack |
| Description | Fixed Tube Rack 40×14mm dia. |