Learn More
Ebro Electronic™ Accessories for EBI 20 Data Logger Series
Click to view available options
Type:
Interface
Mounting Holder
Unit Size:
Each
Description
- Connected to the PC and supplied with power from there by means of a USB cable
- Supplied with Interface, USB cable, software Winlog.basic, instruction manual
- Clean with a slightly damp cloth only
Order Info
Interface
Specifications
Specifications
| Type | Interface |
| Display Type | No Display |
| Calibration | Not Calibrated |
| Alarm Types | None |
| Includes | Interface EBI 20-IF, USB-cable, Software Winlog.basic, instructions for use. |
| Interface | EBI 20-IF |
| Model | EBI 20-IF |
| Sensor Types | External |
Product Content Correction
Product Title
Ebro Electronic™ Accessories for EBI 20 Data Logger Series >