Learn More
Thermo Scientific™ Accessories for Digital Heating Shaking Baths
Product Code. p-6097462 Shop All Thermo Scientific Products
Click to view available options
No. of Places:
6
15
24
48
96
Tube Size:
0.2mL
0.5mL
12mm diameter
13mm diameter
15mL
50mL
missing translation for 'unitSize'
Each
Product Code. 15218884
missing translation for 'mfr': Thermo Scientific™ 88880125
Only null left
Add to basket
Add to basket
Use Thermo Scientific Accessories for Digital Shaking Baths to optimize flexibility and performance.
Use Thermo Scientific Accessories for Digital Heating Shaking Baths to optimize flexibility and performance.
  • Warranty: 2 years
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.2 mL
For Use With (Equipment) Heating/Cooling Bath
Lighting LED
No. of Places 96
Type Block Set with Cover
Tube Size 0.2mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.