Learn More
Merck Accessories for All-glass Filter Holder Kits, 90 mm
Accessories for filter holders for sample preparation.
Brand: Merck XX1019003
3855.59 NOK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.86410kg
Description
Specifications
Not Autoclavable | |
90 mm all-glass filter holder | |
Non-sterile | |
90 mm |
Spring Clamp | |
Anodized Aluminum | |
Spring Clamp, Aluminum |