Description
- Access Port
- With Silicone Plug
- For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Specifications
| Description | Access port with silicone plug - 30 mm, right |
| For Use With (Equipment) | BINDER Series CB and CBF |
| Material | Silicone |
| Type | Access Port |
| Features | Right Position |
| Includes | Silicone plug |
| Diameter (English) | 1.2 in. |
| Diameter (Metric) | 30 mm |