missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Access Port with Silicone Plug for KBW 720 Growth Chambers

This Access Port with Silicone Plug is designed for use with BINDER™ KBW 720 Growth Chambers with Light.

Brand:  BINDER™ 8012-1463

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 16454249

  • 10856.93 NOK / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket

Description

Description

  • Access port with silicone plug
  • Available in assorted sizes and positions
Specifications

Specifications

Access port with silicone plug - 100 mm, top
Silicone
BINDER™ KBW Growth Chambers with Light
100 mm
BINDER Chambers Series KBW
KBW 720
3.94 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers