Learn More
Thermo Scientific ACCESS PORT LEFT DIA.50MMthe middle of the left side wall D = 58 mm
Brand:  Thermo Scientific 51900997
ACCESS PORT LEFT DIA.50MMthe middle of the left side wall D = 58 mm
Product Code. 11804423
  • ONLINE EXCLUSIVE PRICE
    5815.00 NOK
    5510.13 NOK / Each
    Save 304.87 NOK
    5% Off
Only null left
Estimated Shipment: 10-03-2026