Learn More
BINDER™ Access Port for BINDER™ KB ECO Series
Product Code. 17552875 Shop All BINDER Products
Click to view available options
Port Size:
100 mm Left
100 mm Right
100 mm Top
30 mm Left
30 mm Right
30 mm Top
50 mm Left
50 mm Right
50 mm Top
Unit Size:
Each
Product Code. 17552875
Brand: BINDER™ 80122112
Only null left
Add to basket
Add to basket
The Access Port for BINDER KB ECO Series is an option for Series KB ECO Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
  • Access port with silicone plug
Specifications
Description Access port with silicone plug - 30 mm, right
For Use With (Equipment) BINDER Chambers Series KB ECO
Material Steel
Type Access Port
Port Size 30 mm Right