Learn More
BINDER™ Access Port
Product Code. 30238375 Shop All BINDER Products
Change view
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
Unit Size:
Each
Product Code. Port Size Ports unitSize
30238375 100 mm 1 on Left Side Each
30238232 30 mm 1 on Left Side Each
30238330 50 mm 1 on Left Side Each
30238344 30 mm 1 on Right Side Each
30238259 50 mm 1 on Right Side Each
30238256 100 mm 1 on Right Side Each
30238361 50 mm 1 on Top Each
30238210 100 mm 1 on Top Each
30238332 30 mm 1 on Top Each
9 options
Product Code. 30238375
Brand: BINDER™ 80122483
Only null left
Add to basket
Add to basket
Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600
Specifications
For Use With (Equipment) KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Material Silicone
Type Access Port
Ports 1 on Left Side
Port Size 100 mm