Learn More
BINDER™ Access Port
Product Code. 30238257 Shop All BINDER Products
Change view
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
Unit Size:
Each
Product Code. Port Size Ports unitSize
30238257 100 mm 1 on Left Side Each
30238338 30 mm 1 on Left Side Each
30238219 50 mm 1 on Left Side Each
30238239 30 mm 1 on Right Side Each
30238382 50 mm 1 on Right Side Each
30238290 100 mm 1 on Right Side Each
30238370 50 mm 1 on Top Each
30238301 100 mm 1 on Top Each
30238355 30 mm 1 on Top Each
9 options
Product Code. 30238257
Brand: BINDER™ 80122475
Only null left
Add to basket
Add to basket
Access port with silicone plug for use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600.
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600
Specifications
For Use With (Equipment) KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600
Material Silicone
Type Access Port
Ports 1 on Left Side
Port Size 100 mm