Learn More
BINDER™ Access Port
Product Code. 30238203 Shop All BINDER Products
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
Unit Size:
Each
Product Code. 30238203
Brand: BINDER™ 80122472
Only null left
Add to basket
Add to basket
Access port with silicone plug for use with KB65, KB130, KB260, KB470, KB720, and KB 1060.
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KB65, KB130, KB260, KB470, KB720, and KB 1060
Specifications
For Use With (Equipment) KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060
Material Silicone
Type Access Port
Ports 1 on Top
Port Size 100 mm