Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Shop All Thermo Scientific ProductsDescription
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
- Card access comes with 5 key cards
- Field installed - purchased separately
- Factory installed in the plant with purchase of unit
Specifications
Specifications
| Type | Factory-installed. No stand-alone orders. |
| Temperature Range | -40°C to -86C°C |
| No. of Doors | 2 |
| Shelves | 3 Adjustable |
| Style | Upright |
| For Use With (Equipment) | TSX Universal (-ULTS Models) |
| Includes | Access key option; Card access w/five key cards. All orders must accompany a TSX Universal ultra-low freezer |
| Monitoring Options | Audible and Visual High and Low Temperature Alarms |
| Voltage | 100/240 V |
| Compressor | Variable Speed |