Learn More
Acarbose D-Fructose Impurity, TRC
Specifications
Specifications
| Analyte or Component Names | O-4,6-Dideoxy-4-[{(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-enyl}amino]-alpha-D-glucopyranosyl-(1-4)-O-alpha-D-glucopyranosyl-(1-4)-D-arabino-hex-2-ulopyranose |
| Shipping Condition | Room Temperature |
| InChI Formula | InChI=1S/C25H43NO18/c1-7-12(26-9-2-8(3-27)13(31)16(34)14(9)32)15(33)18(36)23(41-7)44-21-11(4-28)42-24(19(37)17(21)35)43-20-10(30)5-40-25(39,6-29)22(20)38/h2,7,9-24,26-39H,3-6H2,1H3/t7-,9+,10-,11-,12-,13-,14+,15+,16+,17-,18-,19-,20-,21-,22+,23-,24-,25?/m1/s1 |
| Solution Type | Neat |
| Industry Type | Forensics |
| Quantity | 1 mg |
| Formula Weight | 645.248 |
| Chemical Name or Material | O-4,6-Dideoxy-4-[{(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-enyl}amino]-alpha-D-glucopyranosyl-(1-4)-O-alpha-D-glucopyranosyl-(1-4)-D-arabino-hex-2-ulopyranose |