Learn More
MedChemExpress Ac-Leu-Arg-AMC
Product Code. 16409548
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
16409548 10 mg 10mg
1 options
Product Code. 16409548
Brand: MedChemExpress HYP1448
MedChemExpress Ac-Leu-Arg-AMC is a fluorogenic peptide substrate.
Specifications
CAS 929621-79-4
Solubility DMSO : 300 mg/mL (616.57 mM∣Need ultrasonic and warming)
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Neuroscience-Neuromodulation
Quantity 10 mg
Molecular Weight (g/mol) 486.56 g/mol
Product Type Dye Reagents
Purity or Quality Grade 99.28%
Shipping Condition Room temperature