Learn More
MedChemExpress Ac-Leu-Arg-AMC
Specifications
Specifications
| CAS | 929621-79-4 |
| Solubility | DMSO : 300 mg/mL (616.57 mM∣Need ultrasonic and warming) |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 486.56 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 99.28% |
| Shipping Condition | Room temperature |