missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AC-55541, MedChemExpress
Description
AC-55541 is a highly selective protease-activated receptor 2 (PAR2) agonist (pEC50=6.7), displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays and exhibits pronociceptive activity in vivo.
Specifications
Specifications
| Chemical Name or Material | AC-55541 |
| CAS | 916170-19-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H20BrN5O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 51 mg/mL (98.39 mM) |
| SMILES | O=C(NC(C(C1=CC=CC=C12)=NNC2=O)C(N/N=C(C)/C3=CC=CC(Br)=C3)=O)C4=CC=CC=C4 |
| Molecular Weight (g/mol) | 518.36 |
| Formula Weight | 518.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?