Learn More
Testo™ 635-2 Temperature and Moisture Meter
Description
- Optional sensors for moisture / temperature / pressure dew point / absolute pressure measurements
- 2 plug-in and 3 wireless sensors
- Super-size memory for up to 10,000 readings
- USB cable
- PC software for logging and archiving measuring data
Specifications
Specifications
| Humidity Range | 0 to 100% |
| Resolution | 0.1%rH |
| Accuracy | NTC: ±0.2°C (−25 - 74.9°C), K TC: ±0.3°C (−60 - 60°C) |
| Temperature Range (Metric) | -40°C, -150°C (NTC), -200°C to +1370°C (KTC) |
| Material | ABS, TPE, Metal |
| Temperature Resolution | 0.1°C |
| Dimensions (L x W x H) | 220 x 74 x 46 mm |
| Weight (Metric) | 428 g |