Learn More
Multiroir Controlec™ ABS Storage Unit with Removable and Tilt-able Drawers
Product Code. 11764581
Change view
Click to view available options
Dimensions (L x W x H):
600 x 133 x 164 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11764581 600 x 133 x 164 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11764581 Supplier Multiroir Controlec™ Supplier No. 50005
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Access stored products easily with the Multiroir Controlec™ ABS Storage Unit with Removable and Tilt-able Drawers. This unit has a tilting tray with a front opening that opens at 45° and allows easy and fast handling of stored products.
  • Highly resistant ABS material
  • Unit color beige
  • Drawer color: transparent
  • Front opening at 45°
  • Tilting tray with front opening allows easy and fast handling of stored products.
  • Five shock-resistant ABS drawers
Specifications
Color Beige, Transparent
Material ABS
Shape Rectangular
Includes 5 Drawers
Product Type Rack
Dimensions (L x W x H) 600 x 133 x 164 mm
Length (Metric) 600 mm
Width (Metric) 133 mm
Height (Metric) 164 mm
Height (Metric) Interior 105/146 mm