Learn More
Tocris Bioscience™ ABP 688
GREENER_CHOICE
High affinity human mGlu5 receptor antagonist
Brand:  Tocris Bioscience™ 5229/10
Encompass
Product Code. 15726596
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ABP 688 is a high affinity human mGlu5 receptor antagonist (Ki = 1.7 nM). Inhibits glutamate-induced calcium release from L(tk-) cells expressing human mGlu5 receptors (IC50 = 2.3 nM).
Specifications
Specifications
3-[2-(6-Methyl-2-pyridinyl)ethynyl]-2-cyclohexene-1-one O-methyloxime
924298-51-1