Learn More
Tocris Bioscience™ ABP 688
High affinity human mGlu5 receptor antagonist
Brand: Tocris Bioscience™ 5229/50
This item is not returnable.
View return policy
Description
ABP 688 is a high affinity human mGlu5 receptor antagonist (Ki = 1.7 nM). Inhibits glutamate-induced calcium release from L(tk-) cells expressing human mGlu5 receptors (IC50 = 2.3 nM).Specifications
| 3-[2-(6-Methyl-2-pyridinyl)ethynyl]-2-cyclohexene-1-one O-methyloxime | |
| 924298-51-1 | |
| 50 mg |