missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ABL1, His Tag Recombinant Protein
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase.
Brand: Invitrogen™ PR4348B
Additional Details : Weight : 0.01000kg
Description
The ABL1 has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Specifications
NP_005148.2 | |
50mM Tris, pH 7.5, 100mM NaCl, 0.1mM EDTA, 0.05% NP-40, 5mM DTT, 50% glycerol |
|
100μg | |
RUO - research use only | |
ABL1, ABL, c-ABL | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
125.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
ABL Kinase Family | |
Recombinant | |
Baculovirus |