missing translation for 'onlineSavingsMsg'
Learn More

ABHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26091-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16874615

  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions.
Specifications

Specifications

ABHD1
Polyclonal
Unconjugated
ABHD1
ABHD1, LABH1, Lung alpha/beta hydrolase 1
Rabbit
Antigen Affinity Chromatography
RUO
57742, 84696
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96SE0, Q9QZC8
ABHD1
ABHD1 Fusion Protein Ag23138
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers