Learn More
Thermo Scientific™ Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
GREENER_CHOICE
Product Code. 11514764
Change view
Click to view available options
Color:
Black
Natural
Volume (Metric) Well:
55 μL
120 μL
250 μL
300 μL
Well Shape:
Round, Conical Bottom (V-well)
Square, Conical Bottom (V-well)
Fit for Purpose:
PCR Ready
Unit Size:
Pack of 50
Product Code. Color Volume (Metric) Well Well Shape Fit for Purpose unitSize
11514764 Natural 300 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10701534 Natural 120 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10324663 Black 120 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10602674 Natural 250 μL Square, Conical Bottom (V-well) PCR Ready Pack of 50
10532255 Natural 55 μL Round, Conical Bottom (V-well) PCR Ready Pack of 50
5 options
Product Code. 11514764
Brand: Thermo Scientific™ AB0862
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
384-well microarray plate with pyramidal well bottoms for maximum sample recovery.
Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Highlights
  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination
Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.
Certifications
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
Autoclavable Autoclavable
Packaging 48 Pk.
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 384-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 384
Volume (Metric) Working 250 μL
Sterility Cleanroom
Surface Treatment Non-treated
Well Shape Square, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF 5000 x g
No. per Case 50
No. per Pack 50
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard