Learn More
ABCF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10226-1-AP-150UL
 View more versions of this product
Product Code. 16846473
  • 4836.10 NOK / 150µL
Description
OCIAD1 was identified via immunoscreening of an ovarian carcinoma cDNA library from ovarian cancer patients and is expressed in multiple tissues including ovary, placenta, brain, testis, prostate, and mammary gland. Two isoforms of OCIAD1 are known to exist; the shorter isoform is restricted to the central nervous system. OCIAD1 is a transmembrane protein whose overexpression in HEY ovarian cancer cells increased lysophosphatidic acid- (LPA-)induced, but not basal level cell adhesion to extracellular matrix proteins collagen I and laminin10/11. This adhesion is not blocked by LY294002 and GF109203X, suggesting that OCIAD1 does not use protein kinase C and PI3 kinase signaling pathways to exert its effect on adhesion.
Specifications
Specifications
ABCF2
Polyclonal
Unconjugated
ABCF2
ABC28, ABCF2, EST133090, HUSSY 18, HUSSY18, M ABC1
Rabbit
Antigen Affinity Chromatography
RUO
10061, 27407
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99LE6, Q9UG63
Abcf2
ABCF2 Fusion Protein Ag0310
150 μL
Primary
Human, Mouse