missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11158-1-AP-20UL
This item is not returnable.
View return policy
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a half-transporter involved in the transport of heme from the mitochondria to the cytosol. With iron/sulfur cluster precursors as its substrates, this protein may play a role in metal homeostasis. Mutations in this gene have been implicated in X-linked sideroblastic anemia with ataxia.Specifications
| ABCB7 | |
| Polyclonal | |
| Unconjugated | |
| ABCB7 | |
| ABC transporter 7 protein, ABC7, ABCB7, ASAT, Atm1p, EST140535 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11306, 22, 302395 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75027, Q61102, Q704E8 | |
| ABCB7 | |
| ABCB7 Fusion Protein Ag1544 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction