missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114801
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ABCB5.
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules.
Specifications
| ABCB5 | |
| Polyclonal | |
| Unconjugated | |
| ABCB5 | |
| 9230106F14Rik; ABCB5; ABCB5 P-gp; ABCB5alpha; ABCB5beta; ATP binding cassette subfamily B member 5; ATP-binding cassette protein; ATP-binding cassette sub-family B member 5; ATP-binding cassette, subfamily B (MDR/TAP), member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; EST422562; P-glycoprotein ABCB5; RGD1566342 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 314537, 340273, 77706 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| B5X0E4, Q2M3G0 | |
| ABCB5 | |
| A synthesized peptide derived from human ABCB5(Accession Q2M3G0), corresponding to amino acid residues R885-A935. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction