Learn More
ABCB5 Monoclonal Antibody (5H3C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517026
Descrizione
MA5-17026 targets ABCB5 in FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17026 immunogen is purified recombit fragment of human ABCB5 (amino acids: 481-674) expressed in E. Coli. MA5-17026 detects ABCB5 which has a predicted molecular weight of approximately 89.8kDa.
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules
Specifica
| ABCB5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q2M3G0 | |
| Mouse | |
| Purified | |
| RUO | |
| 340273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 5H3C6 | |
| Unconjugated | |
| ABCB5 | |
| ABCB5 | |
| Purified recombit fragment of human ABCB5 (amino acids: 481-674) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.