Learn More
Abcam Rat monoclonal [JC5-1] Anti-Mouse lambda light chain (Alkaline Phosphatase), 500 uL
Product Code. 32109351 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109351 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32109351 Supplier Abcam Supplier No. ab99624500uL
 Restricted Item
Estimated Shipment: 02-12-2026
Rat Monoclonal antibody, suitable for Flow Cyt, ICC/IF, ELISA.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications Flow Cytometry
Classification Monoclonal
Clone JC5-1
Conjugate Unconjugated
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene Iglc2
Gene Symbols Iglc2
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Iglc2, Mouse
Validated Application abrv Flow Cyt, ICC/IF, ELISA